Aluminium ≥99.99% (metals basis), foil, annealed, Thickness 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
040531-GH
040531-HP
040531-FI
AA40531-GHEA
123
USD
AA40531-GH
AA40531-HP
AA40531-FI
Aluminium ≥99.99% (metals basis), foil, annealed, Thickness 0.5 mm (0.02 in)
Aluminium
MDL: MFCD00134029
Purity: 99.99%
CAS No.: 7429-90-5
Physical Form: ≈3.4g/50x50mm, Temper: soft
Odor: Odorless
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2327 °C (1013 hPa) Melting Pt: 660 °C Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 Merck Index: 13,00321 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...