Platinum ≥99.95% (metals basis), slug, Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

040419-KG
AA40419-KGEA 862 USD
AA40419-KG
Platinum ≥99.95% (metals basis), slug, Length 6.35 mm (0.25 in), Diameter 6.35 mm (0.25 in)
Platinum

Applications: Evaporation source, jewelry production, electrical contacts for semiconductors


Purity: 99.95% CAS No.: 7440-06-4 Physical Form: ≈4.32g/slug Odor: Odorless MDL No.: MFCD00011179


Evaporation source, jewelry production, electrical contacts for semiconductors

Formula: Pt
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR