Antimony(III) chloride ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Antimony trichloride
040372-04
AA40372-04EA
85.8
USD
AA40372-04
Antimony(III) chloride ≥99.999% (metals basis), Puratronic®
Antimony (III) chloride
MDL: MFCD00011212
Fieser: 8,19 18,25 19,14
Purity: 99.999%
CAS No.: 10025-91-9
Molecular Formula: SbCl3
Formula Weight: 228.11
Physical Form: Crystalline Aggregates
Density: 3.14
Melting Point: 73.4°
Boiling Point: 223.5°
Merck Reference: 14,707
UN No.: 1733
MDL No.: MFCD00011212
Formula:
SbCl₃ Boiling Pt: 223.5 °C Melting Pt: 73.4 °C Density: 3.14 Storage Temperature: Ambient |
MDL Number:
MFCD00011212 CAS Number: 10025-91-9 EINECS: 233-047-2 UN: 1733 ADR: 8,II Merck Index: 14,707 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...