Indium ≥99.999% (metals basis), shot, Puratronic® 1-3 mm (0.04-0.12 in)
Supplier: Thermo Scientific Chemicals
040338-18
040338-30
040338-09
AA40338-18EA
533
USD
AA40338-18
AA40338-30
AA40338-09
Indium ≥99.999% (metals basis), shot, Puratronic® 1-3 mm (0.04-0.12 in)
Indium
MDL: MFCD00134048
Purity: 99.999%
CAS No.: 7440-74-6
Physical Form: Spherical shot
Appearance: Silver
Odor: Odorless
MDL No.: MFCD00134048
Formula:
In MW: 114.82 g/mol Boiling Pt: 2000 Melting Pt: 156 Density: 7.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...