Praseodymium ≥99.5% (metals basis excluding Ta), rods, Diameter 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals

040296-DH 040296-HF 040296-DJ 040296-DM
AA40296-DHEA 573 USD
AA40296-DH AA40296-HF AA40296-DJ AA40296-DM
Praseodymium ≥99.5% (metals basis excluding Ta), rods, Diameter 12.7 mm (0.5 in)
Praseodymium

MDL: MFCD00011174


Purity: 99.5% CAS No.: 7440-10-0 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.86g/mm, Cast rod, diameter is approximate Odor: Odorless

Formula: Pr
MW: 140.91
Storage Temperature: Ambient
MDL Number: MFCD00011174
CAS Number: 7440-10-0
EINECS: 231-120-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR