Lanthanum ≥99.9% (REO, rare earth oxide basis), rods, Diameter 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
040292-DH
040292-DJ
040292-DM
040292-HF
AA40292-DHEA
234
USD
AA40292-DH
AA40292-DJ
AA40292-DM
AA40292-HF
Lanthanum ≥99.9% (REO, rare earth oxide basis), rods, Diameter 6.35 mm (0.25 in)
Lanthanum
MDL: MFCD00011066
Purity: 99.9%
CAS No.: 7439-91-0
Molecular Formula: Packaged in mylar under argon
Physical Form: ≈0.20g/mm, Cast rod, diameter is approximate
Appearance: Gray to silver
UN No.: 3208
MDL No.: MFCD00011066
Formula:
La Boiling Pt: 3464 °C (1013 hPa) Melting Pt: 920 °C Density: ∼6,2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011066 CAS Number: 7439-91-0 EINECS: 231-099-0 UN: 3208 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...