Gadolinium ≥99.9% (metals basis excluding Ta), rods, Diameter 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals

040290-DH 040290-HF
AA40290-DHEA 704 USD
AA40290-DH AA40290-HF
Gadolinium ≥99.9% (metals basis excluding Ta), rods, Diameter 12.7 mm (0.5 in)
Gadolinium

MDL: MFCD00011022


Purity: 99.9% CAS No.: 7440-54-2 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.97g/mm, Cast rod, diameter is approximate MDL No.: MFCD00011022

Formula: Gd
MW: 157.25
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR