Silver(II) oxide ≥99.9% (metals basis) Ag ≥ 86.6%
Supplier: Thermo Scientific Chemicals
|
Danger
|
040187-06
040187-14
040187-22
AA40187-06EA
98.2
USD
AA40187-06
AA40187-14
AA40187-22
Silver(II) oxide ≥99.9% (metals basis) Ag ≥ 86.6%
Silver (II) oxide
MDL: MFCD00044285
Purity: 99.9%
CAS No.: 1301-96-8
Molecular Formula: AgO
Formula Weight: 123.87
Physical Form: -100 Mesh Powder
Density: 7.44
Appearance: Grey to black
Odor: Odorless
Melting Point:
Formula:
AgO MW: 123.87 Melting Pt: 100 (dec.) Density: 7.44 Storage Temperature: Ambient |
MDL Number:
MFCD00044285 CAS Number: 1301-96-8 EINECS: 215-098-2 UN: 3085 ADR: 5.1 II Merck Index: 14,8522 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...