Lead(II) carbonate ACS

Supplier: Thermo Scientific Chemicals
Danger

040119-36 040119-57 040119-22
AA40119-36EA 298.21 USD
AA40119-36 AA40119-57 AA40119-22
Lead(II) carbonate ACS
Lead (II) carbonate

MDL: MFCD00064631


CAS No.: 598-63-0 Molecular Formula: PbCO3 Formula Weight: 267.20 Physical Form: Powder Odor: Odorless UN No.: 2291 MDL No.: MFCD00064631

Formula: PbCO₃
Melting Pt: 315 °C
Density: 6.43 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064631
CAS Number: 598-63-0
EINECS: 209-943-4
UN: 2291
ADR: 6.1,III

Order Now

Specification Test Results

Insoluble in dilute acetic acid 0.02%
Cl 0.2%
Nitrate and nitrite (as NO3) Pass test
Ca 0.01%
K 0.02%
NA 0.05%
Cd 0.002%
Fe 0.005%
Zn 0.003%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR