di-Sodium tetraborate decahydrate 99.5-105.0% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Borax tetra, Sodium tetraborate decahydrate, Sodium Borate decahydrate
040114-36
AA40114-36EA
67.86
USD
AA40114-36
di-Sodium tetraborate decahydrate 99.5-105.0% ACS
di-Sodium tetraborate decahydrate
MDL: MFCD00149193
Notes: 100°C -5H2O, 150°C -9H2O, 320°C -10H2O
Soluble in water, glycerol. Insoluble in alcohol
Purity: 99.5-105.0%
CAS No.: 1303-96-4
Molecular Formula: B4Na2O7.10H2O
Formula Weight: 381.37 (201.22anhy)
Physical Form: Powder
Density: 1.73
Appearance: Grey, blue, or green
Odor: Odorless
pH: 9,24
Melting Point: 75°
Solubility: Soluble in water, glycerol. Insoluble in alcohol
Merck Reference: 14,8590
MDL No.: MFCD00149193
Formula:
Na₂B₄O₇·10H₂O MW: 381.37 g/mol Boiling Pt: 320 °C (1013 hPa) Melting Pt: 75 °C Density: 1.73 Storage Temperature: Ambient |
MDL Number:
MFCD00149193 CAS Number: 1303-96-4 EINECS: 215-540-4 REACH: 01-2119490790-32 Merck Index: 14,8590 |
Specification Test Results
Insoluble matter | 0.005% |
Cl | 0.001% |
PO3 | 0.001% |
SO3 | 0.005% |
Ca | 0.005% |
Heavy Metals (as Pb) | 0.001% |
Fe | 5 ppm |
pH of a 0.01M solution | 9.15-9.20 at 25° |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...