Gold ≥99.96% (metals basis), spherical powder APS 1,5-3,0 µm

Supplier: Thermo Scientific Chemicals

039818-03 039818-06 039818-MD
AA39818-03EA 287 USD
AA39818-03 AA39818-06 AA39818-MD
Gold ≥99.96% (metals basis), spherical powder APS 1,5-3,0 µm
Gold

MDL: MFCD00003436 Notes: Apparent density 40-70g/in3, Tap density: 4.0-9.0g/cm3


Purity: 99.96+% CAS No.: 7440-57-5 Physical Form: S.A. 0.10-0.35m2/g MDL No.: MFCD00003436

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR