Aluminium oxide, γ-phase ≥99.97% (metals basis)
Supplier: Thermo Scientific Chemicals
039812-14
039812-22
039812-36
AA39812-14EA
50.8
USD
AA39812-14
AA39812-22
AA39812-36
Aluminium oxide, γ-phase ≥99.97% (metals basis)
Aluminium oxide, γ-phase
MDL: MFCD00003424
Purity: 99.97%
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: 3 Micron APS Powder, S.A. 80-120m2/g
Odor: Odorless
Merck Reference: 14,356
MDL No.: MFCD00003424
Formula:
Al₂O₃ Boiling Pt: 2980 °C (1013 hPa) Melting Pt: 2045 °C Density: 3.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 Merck Index: 14,356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...