Crucibles, Cylindrical, Graphite, Thermo Scientific
Supplier: Thermo Scientific Chemicals
039811-KN
AA39811-KNEA 169 USD
AA39811-KN
Crucibles, Cylindrical, Graphite, Thermo Scientific
Crucibles
The ultra carbon division graphite crucibles are made from materials exclusively designed for the analysing of gases in metals.
Physical Form: Ultra "F" purity, For O2-H2 analysis