Crystal Polishing Kit, Thermo Scientific
Supplier: Thermo Scientific Chemicals
039798-KT
AA39798-KTEA 697 USD
AA39798-KT
Crystal Polishing Kit, Thermo Scientific
Polishers and Tumblers
The Crystal Polishing Kit contains everything needed by the spectroscopist for repolishing and buffing most infrared transmission crystals, including KRS-5
Two ground glass discs are provided – one for fine grinding and the other to which self-stick polishing pads may be affixed.
Delivery information: Kit contains silicon carbide, 400 and 600 grinding compound; Cerium oxide polishing compound; Aluminium oxide polishing compound (ideal for resurfacing KRS-5 crystals); Finger cots; 3× Camel hair brushes; 2× Polishing pads; 2× Ground glass plates; 5× Polyethylene squeeze bottles with the above-mentioned compounds contained therein; Fitted mahogany base, and instructions.