Potassium bromide for spectroscopy
Supplier: Thermo Scientific Chemicals
|
Warning
|
039796-P2
039796-Q4
AA39796-P2EA
1107
USD
AA39796-P2
AAA39796-Q4
Potassium bromide for spectroscopy
Potassium bromide
MDL: MFCD00011358
Fieser: 21,358
CAS No.: 7758-02-3
Molecular Formula: KBr
Formula Weight: 119.01
Physical Form: Random Cuttings, Chunks
Density: 2.75
Refractive Index: 1.559
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 730°
Boiling Point: 1435°
Merck Reference: 14,7618
MDL No.: MFCD00011358
Formula:
KBr MW: 119 g/mol Boiling Pt: 1435 Melting Pt: 730 Density: 2.75 Storage Temperature: Ambient |
MDL Number:
MFCD00011358 CAS Number: 7758-02-3 EINECS: 231-830-3 Merck Index: 14,7618 |
Specification Test Results
Form | Random Cuttings, Chunks |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...