Tungsten ≥99.9% (metals basis), Powder -325 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
039749-22
039749-36
039749-A3
AA39749-22EA
64.51
USD
AA39749-22
AA39749-36
AA39749-A3
Tungsten ≥99.9% (metals basis), Powder -325 mesh
Tungsten
MDL: MFCD00011461
Purity: 99.9%
CAS No.: 7440-33-7
Appearance: Grey
UN No.: 3089
MDL No.: MFCD00011461
Formula:
W Storage Temperature: Ambient |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 UN: 3089 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...