Tungsten ≥99.9% (metals basis), Powder -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

039749-22 039749-36 039749-A3
AA39749-22EA 64.51 USD
AA39749-22 AA39749-36 AA39749-A3
Tungsten ≥99.9% (metals basis), Powder -325 mesh
Tungsten

MDL: MFCD00011461


Purity: 99.9% CAS No.: 7440-33-7 Appearance: Grey UN No.: 3089 MDL No.: MFCD00011461

Formula: W
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
UN: 3089
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR