Gold(III) acetate ≥99.9% (metals basis)
Supplier: Thermo Scientific Chemicals
039742-02
039742-03
039742-06
AA39742-02EA
160
USD
AA39742-02
AA39742-03
AA39742-06
Gold(III) acetate ≥99.9% (metals basis)
Gold (III) acetate
Purity: 99.9%
CAS No.: 15804-32-7
Molecular Formula: Au(O2CCH3)3
Formula Weight: 374.10
Physical Form: Powder
Formula:
C₆H₉O₆Yb MW: 374.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00210614 CAS Number: 15804-32-7 EINECS: 244-137-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...