Cobalt ≥99.9% (metals basis), foil 50x50 mm (2x2 in), Thickness 0.127 mm (0.005 in)

Supplier: Thermo Scientific Chemicals
Danger

039715-KJ 039715-KG
AA39715-KJEA 677 USD
AA39715-KJ AA39715-KG
Cobalt ≥99.9% (metals basis), foil 50x50 mm (2x2 in), Thickness 0.127 mm (0.005 in)
Cobalt

MDL: MFCD00010935


Purity: 99.9% CAS No.: 7440-48-4 Physical Form: ≈2.83g/50x50mm Odor: Odorless MDL No.: MFCD00010935

Formula: Co
Boiling Pt: 2870 °C (1013 hPa)
Melting Pt: 1495 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR