Zinc ≥99.9% (metals basis), powder -140+325 mesh

Supplier: Thermo Scientific Chemicals
Danger

039694-22 039694-36
AA39694-22EA 211.11 USD
AA39694-22 AA39694-36
Zinc ≥99.9% (metals basis), powder -140+325 mesh
Zinc

MDL: MFCD00011291


Purity: 99.9% CAS No.: 7440-66-6 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: Gray, bluish-white Odor: Odorless UN No.: 1436 MDL No.: MFCD00011291

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
UN: 1436
ADR: 4.3 III
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR