Vanadium naphthenate oxide 35% in naphthenic acid (V 2.8-3.2%)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Vanadyl naphthenate
039585-30
AA39585-30EA
694
USD
AA39585-30
Vanadium naphthenate oxide 35% in naphthenic acid (V 2.8-3.2%)
Vanadium naphthenate oxide
CAS No.: 68553-60-6
Physical Form: Liquid
Density: 1.04
UN No.: 2810
MDL No.: MFCD00148763
Formula:
C₄₄H₂₈O₈V Density: 1.04 Storage Temperature: Ambient |
MDL Number:
MFCD00148763 CAS Number: 68553-60-6 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...