Potassium manganate
Supplier: Thermo Scientific Chemicals
|
Danger
|
039506-09
039506-18
AA39506-09EA
87.5
USD
AA39506-09
AA39506-18
Potassium manganate
Potassium manganate
MDL: MFCD00043089
Soluble in water
CAS No.: 10294-64-1
Molecular Formula: K2MnO4
Formula Weight: 197.13
Physical Form: Powder
Appearance: Dark green
Melting Point: 190° dec.
UN No.: 1479
MDL No.: MFCD00043089
Formula:
K₂MnO₄ Melting Pt: 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00043089 CAS Number: 10294-64-1 EINECS: 233-665-2 UN: 1479 ADR: 5.1,III Merck Index: 14,7644 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...