Potassium manganate

Supplier: Thermo Scientific Chemicals
Danger

039506-09 039506-18
AA39506-09EA 87.5 USD
AA39506-09 AA39506-18
Potassium manganate
Potassium manganate

MDL: MFCD00043089 Soluble in water


CAS No.: 10294-64-1 Molecular Formula: K2MnO4 Formula Weight: 197.13 Physical Form: Powder Appearance: Dark green Melting Point: 190° dec. UN No.: 1479 MDL No.: MFCD00043089

Formula: K₂MnO₄
Melting Pt: 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00043089
CAS Number: 10294-64-1
EINECS: 233-665-2
UN: 1479
ADR: 5.1,III
Merck Index: 14,7644

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR