Barium tetracyanoplatinate(II) tetrahydrate ≥99.99% (metals basis), Premion®

Supplier: Thermo Scientific Chemicals
Danger

039463-03
AA39463-03EA 318 USD
AA39463-03 AA39463-06
Barium tetracyanoplatinate(II) tetrahydrate ≥99.99% (metals basis), Premion®
Barium tetracyanoplatinate(II) tetrahydrate

Purity: 99.99% CAS No.: 13755-32-3 Molecular Formula: BaPt(CN)4.4H2O Formula Weight: 508.56 (436.50anhy) Physical Form: Powder Density: 3.05 Odor: Odorless Melting Point: 100° dec. UN No.: 1564 MDL No.: MFCD00014183

Formula: BaPt(CN)₄.4H₂O
Melting Pt: 100 °C (dec.)
Density: 3.05
Storage Temperature: Ambient
MDL Number: MFCD00014183
CAS Number: 13755-32-3
EINECS: 209-238-1
UN: 1564
ADR: 6.1,II
Merck Index: 14,991

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR