Dihydrogen hexafluorozirconate 20% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zirconate(2-), hexafluoro-, Hexafluorozirconic acid, Hexafluorozirconate(2-)

039461-A1
AA39461-A1EA 74.4 USD
AA39461-A1 AA39461-A7
Dihydrogen hexafluorozirconate 20% (w/w) in aqueous solution
Dihydrogen hexafluorozirconate

CAS No.: 12021-95-3 Molecular Formula: H2ZrF6 Formula Weight: 207.22 Physical Form: Liquid Density: 1.52 Appearance: Pale green UN No.: 3264 MDL No.: MFCD00082965

Formula: H₂ZrF₆
Density: 1.52
Storage Temperature: Ambient
MDL Number: MFCD00082965
CAS Number: 12021-95-3
EINECS: 234-666-0
UN: 3264
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR