Calcium stearate

Supplier: Thermo Scientific Chemicals

Synonyms: Stearic acid calcium salt

039423-A1 039423-A7
AA39423-A1EA 73.4 USD
AA39423-A1 AA39423-A7
Calcium stearate
Calcium stearate

Waterproofing and release agent, stabilizer for PVC, lubricant, conditioning agent


CAS No.: 1592-23-0 Molecular Formula: Ca[CH3(CH2)16CO2]2 Formula Weight: 607.04 Physical Form: Powder Odor: Odorless Melting Point: 179-180° MDL No.: MFCD00036390


Waterproofing and release agent, stabilizer for PVC, lubricant, conditioning agent

Formula: C₃₆H₇₀CaO₄
Melting Pt: 179…180 °C
Density: 1.065 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036390
CAS Number: 1592-23-0
EINECS: 216-472-8
Merck Index: 14,1703
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66064c431fc65dcdf98ff2bc1d0227f37de78108&vid=c1765b9467ae214d145f3824443319aa43d8bd1e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR