Copper aluminum oxide ≥99.5%

Supplier: Thermo Scientific Chemicals

039419-30 039419-18
AA39419-30EA 442 USD
AA39419-30 AA39419-18
Copper aluminum oxide ≥99.5%
Copper aluminum oxide

Purity: 99.5% CAS No.: 12042-92-1 Molecular Formula: CuAl2O4, may contain some CuO Formula Weight: 181.50 Physical Form: -325 Mesh Powder MDL No.: MFCD00210610

Formula: CuAl₂O₄, may contain some CuO
Storage Temperature: Ambient
MDL Number: MFCD00210610
CAS Number: 12042-92-1
EINECS: 234-934-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR