Copper aluminum oxide ≥99.5%
Supplier: Thermo Scientific Chemicals
039419-30
039419-18
AA39419-30EA
442
USD
AA39419-30
AA39419-18
Copper aluminum oxide ≥99.5%
Copper aluminum oxide
Purity: 99.5%
CAS No.: 12042-92-1
Molecular Formula: CuAl2O4, may contain some CuO
Formula Weight: 181.50
Physical Form: -325 Mesh Powder
MDL No.: MFCD00210610
Formula:
CuAl₂O₄, may contain some CuO Storage Temperature: Ambient |
MDL Number:
MFCD00210610 CAS Number: 12042-92-1 EINECS: 234-934-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...