Bis(pentamethylcyclopentadienyl)iron(II) 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(pentamethylcyclopentadienyl)iron, Decamethylferrocene

039406-06 039406-MD 039406-03
AA39406-06EA 936 USD
AA39406-06 AA39406-MD AA39406-03
Bis(pentamethylcyclopentadienyl)iron(II) 99%
Bis(pentamethylcyclopentadienyl)iron (II)

Purity: 99% CAS No.: 12126-50-0 Molecular Formula: C20H30Fe Formula Weight: 326.31 Physical Form: Crystalline Sensitivity: Air and Moisture Sensitive MDL No.: MFCD00058736

Formula: C₂₀H₃₀Fe
Melting Pt: 298…300 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00058736
CAS Number: 12126-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR