Bis(pentamethylcyclopentadienyl)iron(II) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bis(pentamethylcyclopentadienyl)iron, Decamethylferrocene
039406-06
039406-MD
039406-03
AA39406-06EA
936
USD
AA39406-06
AA39406-MD
AA39406-03
Bis(pentamethylcyclopentadienyl)iron(II) 99%
Bis(pentamethylcyclopentadienyl)iron (II)
Purity: 99%
CAS No.: 12126-50-0
Molecular Formula: C20H30Fe
Formula Weight: 326.31
Physical Form: Crystalline
Sensitivity: Air and Moisture Sensitive
MDL No.: MFCD00058736
Formula:
C₂₀H₃₀Fe Melting Pt: 298…300 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00058736 CAS Number: 12126-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...