Caffeine ≥99.7%
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00005758
Beilstein Registry No.: 17705
Purity: 99.7%
CAS No.: 58-08-2
Molecular Formula: C8H10N4O2
Formula Weight: 194.19
Physical Form: Powder
Density: 1.230
Appearance: White
Melting Point: 234-236°
Boiling Point: 178° subl.
Beilstein Reference: 17705
Merck Reference: 14,1636
UN No.: 1544
MDL No.: MFCD00005758
Formula:
C₈H₁₀N₄O₂ Boiling Pt: 178 °C (subl.) Melting Pt: 234…236 °C Density: 1.230 Storage Temperature: Ambient |
MDL Number:
MFCD00005758 CAS Number: 58-08-2 EINECS: 200-362-1 UN: 1544 ADR: 6.1,III Merck Index: 14,1636 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...