Butyl acetate ≥99.5% for HPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate
039197-AK
039197-M1
039197-K2
039197-K7
AA39197-AKEA
60.11
USD
AA39197-AK
AA39197-M1
AA39197-K2
AA39197-K7
Butyl acetate ≥99.5% for HPLC
Butyl acetate
MDL: MFCD00009445
Beilstein Registry No.: 1741921
Purity: 99.5+%
CAS No.: 123-86-4
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 25°(77°F)
Physical Form: Liquid
Density: 0.881
Refractive Index: 1.3945
Appearance: Colorless
Odor: Fruit-like
pH: 5
Melting Point: -78°
Boiling Point: 124-126°
Beilstein Reference: 1741921
Merck Reference: 14,1535
UN No.: 1123
MDL No.: MFCD00009445
Formula:
CH₃COO(CH₂)₃CH₃ MW: 116.16 g/mol Boiling Pt: 126 Melting Pt: -76 Density: 0.881 Flash Pt: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009445 CAS Number: 123-86-4 EINECS: 204-658-1 UN: 1123 ADR: 3 III Merck Index: 14,1535 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...