Butyl acetate ≥99.5% for HPLC

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate

039197-AK 039197-M1 039197-K2 039197-K7
AA39197-AKEA 60.11 USD
AA39197-AK AA39197-M1 AA39197-K2 AA39197-K7
Butyl acetate ≥99.5% for HPLC
Butyl acetate

MDL: MFCD00009445 Beilstein Registry No.: 1741921


Purity: 99.5+% CAS No.: 123-86-4 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 25°(77°F) Physical Form: Liquid Density: 0.881 Refractive Index: 1.3945 Appearance: Colorless Odor: Fruit-like pH: 5 Melting Point: -78° Boiling Point: 124-126° Beilstein Reference: 1741921 Merck Reference: 14,1535 UN No.: 1123 MDL No.: MFCD00009445

Formula: CH₃COO(CH₂)₃CH₃
MW: 116.16 g/mol
Boiling Pt: 126
Melting Pt: -76
Density: 0.881
Flash Pt: 25 °C (77 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009445
CAS Number: 123-86-4
EINECS: 204-658-1
UN: 1123
ADR: 3 III
Merck Index: 14,1535
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403329de943f8cb2ea6cd9d73ee35957d2140aa7&vid=682ab82de177b2985e94089408f7efebde59926d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR