Magnesium nitrate solution 1% in nitric acid 5%, Specpure® matrix modifier solution

Supplier: Thermo Scientific Chemicals
Danger

039044-AE
AA39044-AEEA 116.76 USD
AA39044-AE
Magnesium nitrate solution 1% in nitric acid 5%, Specpure® matrix modifier solution
Magnesium nitrate solutions

Molecular Formula: 1% Mg(NO3)2 in 2% HNO3 Physical Form: Liquid UN No.: 3264 MDL No.: MFCD00011103

Formula: Mg(NO₃)₂
Storage Temperature: Ambient
MDL Number: MFCD00011103
CAS Number: 10377-60-3
EINECS: 231-714-2
UN: 3264
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR