Nickel (II) nitrate 1% Ni(NO3)2 in nitric acid 2%, Specpure®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nickel dinitrate

039043-AE
AA39043-AEEA 108 USD
AA39043-AE
Nickel (II) nitrate 1% Ni(NO3)2 in nitric acid 2%, Specpure®
Nickel (II) nitrate

Matrix Modifier Solution

Formula: N₂NiO₆
MW: 182.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011139
CAS Number: 13138-45-9
UN: 3264
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR