Sulfuric acid 93-98%, Environmental Grade

Supplier: Thermo Scientific Chemicals
Danger

038751-K4 038751-AP
AA38751-K4EA 340 USD
AA38751-K4 AA38751-AP
Sulfuric acid 93-98%, Environmental Grade
Sulfuric acid

MDL: MFCD00064589 Notes: Certificate of Analysis included


CAS No.: 7664-93-9 Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid, single sub-boiling quartz distillation Density: 1.84 Appearance: Colorless to pale yellow Odor: Acrid Merck Reference: 14,8974 UN No.: 1830 MDL No.: MFCD00064589

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
Merck Index: 14,8974

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR