Hydrofluoric acid 47 - 51%, Environmental Grade Plus

Supplier: Thermo Scientific Chemicals
Danger

038746-AP
AA38746-APEA 1404 USD
AA38746-AP
Hydrofluoric acid 47 - 51%, Environmental Grade Plus
Hydrofluoric acid

MDL: MFCD00011346 Notes: Special handling precautions required. View MSDS prior to purchase. Certificate of Analysis included


CAS No.: 7664-39-3 Molecular Formula: HF Formula Weight: 20.01 Physical Form: Liquid, double sub-boiling quartz distillation Density: 1.16 Appearance: Colorless Odor: Acrid Boiling Point: 112° Merck Reference: 14,4790 UN No.: 1790 MDL No.: MFCD00011346

Formula: HF
MW: 20.01
Boiling Pt: 112 °C
Density: 1.16
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8 II
Merck Index: 14,4790

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR