Phenolphthalein ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
038705-22
038705-36
AA38705-22EA
64.6
USD
AA38705-22
AA38705-36
Phenolphthalein ACS
Phenolphthalein
Indicator
CAS No.: 77-09-8
Molecular Formula: C20H14O4
Formula Weight: 318.33
Physical Form: Powder
Density: 1.299
Melting Point: 258-262°
Beilstein Reference: 284423
Merck Reference: 14,7243
MDL No.: MFCD00005913
Indicator
Formula:
C₂₀H₁₄O₄ Melting Pt: 258…262 °C Density: 1.299 Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 EINECS: 201-004-7 REACH: 01-2119498295-24 Merck Index: 14,7243 |
|
Specification Test Results
Clarity of alcohol solution | Pass test |
Visual transition interval | pH 8.0 (colorless) to 10.0 (red) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...