Aluminium oxide substrate 10x10x0.5 mm, polished one side A plane

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

038498-KT 038498-KU
AA38498-KTEA 85.1 USD
AA38498-KT AA38498-KU
Aluminium oxide substrate 10x10x0.5 mm, polished one side A plane
Aluminium oxide

MDL: MFCD00003424


Molecular Formula: Al2O3 Odor: Odorless MDL No.: MFCD00003424

Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR