Zinc selenide, crystal optic rectangle 38.5 mm x 19.5 mm x 2 mm, polished both sides (drilled)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc (II) selenide

038478-KT
AA38478-KTEA 610 USD
AA38478-KT
Zinc selenide, crystal optic rectangle 38.5 mm x 19.5 mm x 2 mm, polished both sides (drilled)
Zinc selenide
Formula: SeZn
MW: 144.35 g/mol
Melting Pt: 1793 °C
Density: 5.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1,III
Merck Index: 12,10290

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR