Calcium fluoride, crystal optic rectangle 38.5 mm x 19.5 mm x 4 mm, polished both sides (drilled)

Supplier: Thermo Scientific Chemicals
Warning

038476-KT
AA38476-KTEA 256 USD
AA38476-KT
Calcium fluoride, crystal optic rectangle 38.5 mm x 19.5 mm x 4 mm, polished both sides (drilled)
Calcium fluoride

MDL No.: MFCD00010907

Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: 2500 °C (1013 hPa)
Melting Pt: 1418 °C
Density: 3.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 13,01669

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR