Potassium bromide, crystal optic rectangle 38.5 mm x 19.5 mm x 4 mm, polished both sides (drilled)

Supplier: Thermo Scientific Chemicals
Warning

038475-KT
AA38475-KTEA 107 USD
AA38475-KT
Potassium bromide, crystal optic rectangle 38.5 mm x 19.5 mm x 4 mm, polished both sides (drilled)
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435 °C (1013 hPa)
Melting Pt: 730 °C
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR