Barium fluoride, crystal optic rectangle 30 mm x 15 mm x 4 mm, polished both sides

Supplier: Thermo Scientific Chemicals
Warning

038459-KT
AA38459-KTEA 274 USD
AA38459-KT
Barium fluoride, crystal optic rectangle 30 mm x 15 mm x 4 mm, polished both sides
Barium fluoride

Common Applications: Spectroscopy, IR window


Spectroscopy, IR window

Formula: BaF₂
MW: 175.33 g/mol
Boiling Pt: 2260 °C (1013 hPa)
Melting Pt: 1270 °C
Density: 4.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003450
CAS Number: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1,III
Merck Index: 12,01003

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR