Zinc selenide, crystal optic disc 25 mm x 2 mm, polished both sides

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc (II) selenide

038454-KT
AA38454-KTEA 499 USD
AA38454-KT
Zinc selenide, crystal optic disc 25 mm x 2 mm, polished both sides
Zinc selenide
Formula: SeZn
MW: 144.35 g/mol
Melting Pt: 1793 °C
Density: 5.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1,III
Merck Index: 12,10290

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR