Potassium bromide, crystal optic disc (drilled) 32 mm x 3 mm, polished both sides

Supplier: Thermo Scientific Chemicals
Warning

038427-KT
AA38427-KTEA 118 USD
AA38427-KT
Potassium bromide, crystal optic disc (drilled) 32 mm x 3 mm, polished both sides
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435
Melting Pt: 730
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR