Potassium bromide, crystal optic disc 25 mm x 4 mm, polished both sides

Supplier: Thermo Scientific Chemicals
Warning

038425-KT
AA38425-KTEA 131.09 USD
AA38425-KT
Potassium bromide, crystal optic disc 25 mm x 4 mm, polished both sides
Potassium bromide

MDL No.: MFCD00011358

Formula: KBr
Boiling Pt: 1435 °C (1013 hPa)
Melting Pt: 730 °C
Density: 2.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 13,07701

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR