Potassium chloride, crystal optic disc 25 mm x 4 mm, polished both sides

Supplier: Thermo Scientific Chemicals

038420-KT
AA38420-KTEA 79.9 USD
AA38420-KT
Potassium chloride, crystal optic disc 25 mm x 4 mm, polished both sides
Potassium chloride

MDL No.: MFCD00011360

Formula: KCl
Boiling Pt: 1420 °C (1013 hPa)
Melting Pt: 773 °C
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR