Sodium chloride, crystal optic disc 32 mm x 3 mm, polished both sides

Supplier: Thermo Scientific Chemicals

038416-KT
AA38416-KTEA 115.4 USD
AA38416-KT
Sodium chloride, crystal optic disc 32 mm x 3 mm, polished both sides
Sodium chloride
Formula: NaCl
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801 °C
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR