Platinum ≥99.95% (metals basis), foil, Thickness 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

038378-FF
AA38378-FFEA 1152 USD
AA38378-FF
Platinum ≥99.95% (metals basis), foil, Thickness 0.25 mm (0.01 in)
Platinum

MDL: MFCD00011179


Purity: 99.95% CAS No.: 7440-06-4 Physical Form: ≈3.4g/25x25mm Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827 °C (1013 hPa)
Melting Pt: 1769 °C
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR