Platinum 10% on activated carbon wetted with ca. 50% water, powder, unreduced

Supplier: Thermo Scientific Chemicals

038326-06 038326-14 038326-22
AA38326-06EA 137 USD
AA38326-06 AA38326-14 AA38326-22
Platinum 10% on activated carbon wetted with ca. 50% water, powder, unreduced
Platinum

Sold on a dry weight basis. Unit weight excludes water weight


CAS No.: 7440-06-4 Physical Form: 900-1100 m²/g MDL No.: MFCD00011179

Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR